Learn More
Enzo Life Sciences CPN10 (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP110F
Encompass_Preferred
Alternative name: HSP10, HSPE1, GROES, Heat shock protein 10, Chaperonin 10. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~10kDa. Formulation: Liquid. In 20mM TRIS, pH 8.0, containing 150mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: ELISA, WB .
Catalog No. 50-200-7591