Learn More
Aviva Systems Biology Corp CPN2 Antibody - N-terminal region (ARP51877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPN2. Alternate names: ACBPApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60635