Learn More
Aviva Systems Biology Corp CPNE1 Antibody - N-terminal region (ARP40504_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40504_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CPNE1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CPNE1. Alternate names: CPN1, COPN1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57492