Learn More
Aviva Systems Biology Corp CPNE4 Antibody - N-terminal region (ARP52543_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52543_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPNE4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPNE4. Alternate names: COPN4, CPN4, MGC15604ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60766