Learn More
Aviva Systems Biology Corp Cpne7 Antibody - C-terminal region (ARP55052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cpne7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Cpne7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-996