Learn More
Aviva Systems Biology Corp CPNE9 Antibody - middle region (ARP53190_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53190_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPNE9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CPNE9. Alternate names: KIAA4217ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60963