Learn More
Aviva Systems Biology Corp CPNE9 Antibody - C-terminal region (ARP53191_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53191_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPNE9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CPNE9. Alternate names: KIAA4217ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-806