Learn More
Bachem (Cys47)-HIV-1 tat Protein (47-57) Trifluoroacetate
SDP
Supplier:  Bachem 4066788.0005
Encompass_Preferred
(Cys47)-HIV-1 tat Protein (47-57) Trifluoroacetate, H-Cys-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH, 627079-23-6, C58H114N32O13S, Mr 1499.82, 5mg. CPP for gene delivery.
Catalog No. 50-194-6808