Learn More
Medchemexpress LLC Cpp12 (Tfa) | 98.69% | 1700.55 | 1 MG
SDP
Catalog No. 5000425010
Encompass_Preferred
CPP12 TFA is a small, amphiphilic, cyclic cell-penetrating peptide (CPP) in salt form. It binds directly to plasma membrane phospholipids, enters mammalian cells via endocytosis, and is then efficiently released from endosomes. It can be used for intracellular delivery of drugs and chemical probes.
  • Small, amphiphilic, cyclic cell-penetrating peptide (CPP)
  • Binds directly to plasma membrane phospholipids
  • Enters mammalian cells via endocytosis
  • Efficiently released from endosomes
  • Can be used for intracellular delivery of drugs and chemical probes
Catalog No. 50-004-25010 Supplier Medchemexpress LLC Supplier No. HYP10649A1MG
CPP12 TFA is a small, amphiphilic, cyclic cell-penetrating peptide (CPP) in salt form. It binds directly to plasma membrane phospholipids, enters mammalian cells via endocytosis, and is then efficiently released from endosomes. It can be used for intracellular delivery of drugs and chemical probes.
  • Small, amphiphilic, cyclic cell-penetrating peptide (CPP)
  • Binds directly to plasma membrane phospholipids
  • Enters mammalian cells via endocytosis
  • Efficiently released from endosomes
  • Can be used for intracellular delivery of drugs and chemical probes