Learn More
Aviva Systems Biology Corp CPPED1 Antibody - N-terminal region (ARP62267_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62267_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPPED1. It was validated on Western Blot. . Alternate names: CSTP1, FLJ11151ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rat
Catalog No. 50-287-638