Learn More
ZOLL™ CPR Uni-Padz III (Adult/Pediatric) Electrodes
Catalog No. 17300108 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Electrode, CPR Uni-Padz
Catalog No. Product Type
17-300-108 Electrode, CPR Uni-Padz
1 options
Catalog No. 17-300-108 Supplier ZOLL™ Supplier No. 890000028001
Add to Cart
Add to Cart
For adult or child rescue
  • Adult placement standard upper right chest/lower left side
  • Pediatric placement front and back
  • 5 year lifespan
Compatible With:
  • ZOLL™ AED 3
Note:
  • Not approved for use on ZOLL™ AED 3 BLS device
Specifications
Product Type Electrode, CPR Uni-Padz
For Use With (Application) For adult or child rescue
For Use With (Equipment) ZOLL™ AED 3