Learn More
ZOLL™ CPR Uni-Padz III Demonstration/Training Kit
Catalog No. 17300118 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Training Uni-Padz
Catalog No. Product Type
17-300-118 Training Uni-Padz
1 options
Catalog No. 17-300-118 Supplier ZOLL™ Supplier No. 8900000284
Add to Cart
Add to Cart
Connects to AED 3 defibrillator for training and simulations
Comptabile With:
  • ZOLL™ AED 3
  • ZOLL™ AED 3 BLS
Includes:
  • One set of two reusable training pads
Specifications
Product Type Training Uni-Padz
Includes One set of two reusable training pads
For Use With (Application) For training and simulations
For Use With (Equipment) ZOLL™ AED 3, ZOLL™ AED 3 BLS