Learn More
3B Scientific™ CPRLilly PRO+ without Tablet
SDPSCI_ED
Catalog No. S30514 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30514 1 Ea.
1 options
Catalog No. S30514 Supplier 3B Scientific™ Supplier No. 1022237
Add to Cart
Add to Cart
3B Scientific™ CPRLilly PRO+ enables instructors to measure, monitor and analyze the CPR performance of up to 10 trainees at a time. Helps instructors increase effectiveness of their CPR courses by connecting with an App to follow the CPR performance and deliver objective feedback leading to Quality CPR.
Instrumented directive feedback device provides real-time audio or visual feedback (or both) on these critical CPR skills:
  • Compression depth, rate and fraction
  • Chest recoil
  • Correct hand placement
  • Jaw thrust and head tilt
  • Ventilation volume
Key Training Features:
  • Quality CPR feedback with debriefing history
  • Mouth-to-mouth and mouth-to-nose ventilation
  • Ventilation with face shield, pocket mask and BVM
  • Chest rise with proper ventilation
  • Control of proper airway desobstuction
  • Carotid pulses
  • AED pads placement training capable
  • Free App available on google play and iOS
  • Constant analysis of CPR parameters for up to 10 mannequins simultaneously
  • Individual review possibility in group training for more efficient training
  • Adaptable to different course concepts
  • Simplified student feedback view available with the CPRLilly App
Solid mannequin for effective training:
  • Made in Germany of durable, latex-free material
  • Face masks and lung bags can be changed easily without tools
  • Easy to clean with hygienic airway system
  • Runs quietly with efficient power usage and rechargeable battery
  • Stable WiFi connection and intuitive App interface
  • Practical carrying bag with training mat for comfortable practise
  • Dark skin mannequin available
Specifications
For Use With (Application) Measuring, monitoring and analyzing the CPR performance of up to 10 trainees at a time
Includes 1 CPRLilly PRO+ training mannequin, 1 Tablet, 1 charger and battery, 4 lower face masks (including teeth), 25 throat bags, 12 AED-Pad adapters, 1 carrying bag with training mat
Quantity 1 Ea.