Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Certified Safety CPRotector™ 2000
SDP

Catalog No. 19822043
Change view
Click to view available options
Product Type:
CPR Barrier
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-822-043 CPR Barrier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-822-043 Supplier Certified Safety Supplier No. 216068

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Comprehensive instructions included in each package

Prevents unsafe mouth-to-mouth contact

  • Disposable, single-use mouth-to-mouth barriers provide protection to rescuers from victim's oral secretions
  • One-way valve allows maximum amount of air to be sent to victim while preventing return of air or body fluids to rescuer
  • Cross-section gate prevents tongue blockage
  • Gate can also be used as a bite stick
  • Patented design is made of clear, high-density polypropylene
  • Remains soft and flexible to -10°F (-23°C)
  • Instructions for use printed on unit

Order Info

Available packaged in a light smoke poly bag with a pair of latex gloves or without gloves in a loose-fit, easy-to-open clear overwrap.

Compliance

FDA 510(K) registered

Specifications

Product Type CPR Barrier
Color Clear
Material HDPE
Disposable Single-use
For Use With (Application) Prevents unsafe mouth-to-mouth contact
Includes Comprehensive instructions
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.