Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1373MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: CPS1/1022. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa cells. Liver or Hepatocellular Carcinoma (HCC). Cellular Staining: Finely granular cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CPS1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~165kDa.Chromosome: 2q35. Gene Symbol: CPS1. Gene ID: 1373. Ensemble: ENSG00000021826. Uniprot: XP_011508945.
Catalog No. 50-188-4625