Learn More
Hach Company Hach AS950 Portable Compact Sampler, 12 V, 24 x 575 mL bottles (ASP.CXXXC161XX)
Catalog No. NC2610774 Shop All Hach Company Products
Encompass
The AS950 portable compact sampler is a battery-powered field sampler designed for remote and project-based water sampling. It supports discrete and composite sampling, stores detailed datalogging records, and is built for rugged outdoor use.
  • Battery-powered portable sampler with included 12 V lead-acid battery
  • Twenty-four 575 mL sample bottles for discrete sampling
  • Peristaltic pump with corrosion-resistant Delrin resin construction
  • Programmable datalogging controller stores up to 325,000 records
  • Rugged NEMA 4X/6 housing with lockable top cover
  • Double-walled insulated base accepts ice or gel packs to maintain sample temperature
  • Fits in an 18 in manhole for easy field installation
  • Battery charger sold separately
Catalog No. NC2610774 Supplier Hach Company Supplier No. ASP.CXXXC161XX
Add to Cart
Add to Cart
Request a Quote
The AS950 portable compact sampler is a battery-powered field sampler designed for remote and project-based water sampling. It supports discrete and composite sampling, stores detailed datalogging records, and is built for rugged outdoor use.
  • Battery-powered portable sampler with included 12 V lead-acid battery
  • Twenty-four 575 mL sample bottles for discrete sampling
  • Peristaltic pump with corrosion-resistant Delrin resin construction
  • Programmable datalogging controller stores up to 325,000 records
  • Rugged NEMA 4X/6 housing with lockable top cover
  • Double-walled insulated base accepts ice or gel packs to maintain sample temperature
  • Fits in an 18 in manhole for easy field installation
  • Battery charger sold separately