Learn More
Aviva Systems Biology Corp CPSF3 Antibody - N-terminal region (ARP40840_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40840_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CPSF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPSF3. Alternate names: CPSF, YSH1, CPSF73, CPSF-73ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57636