Learn More
TARGETMOL CHEMICALS INC CPSI-1306 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1731510MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (±)-CPSI-1306 (2-(3-(24-Difluorophenyl)-45-dihydroisoxazol-5-yl)-1-morpholinoethan-1-one) is a macrophage migration inhibitory factor (MIF) antagonist and can be used in studies about non-insulin-dependent diabetes mellitus (NIDDM) and the prevention of the deleterious cutaneous effects of acute and chronic UVB exposure. purity: 97%
Catalog No. 50-258-1106