Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-CPSI 1306, Tocris Bioscience™

Catalog No. 440010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-001-0 Supplier R&D Systems Supplier No. 4400/10
This item is not returnable. View return policy

Reported macrophage migration inhibitory factor (MIF) inhibitor

(±)-CPSI 1306 is reported to be a macrophage inhibitory factor (MIF) inhibitor.

This product is racemic.

Chemical Identifiers

CAS 1309793-47-2
Molecular Formula C15H16F2N2O3
Molecular Weight (g/mol) 310.301
InChI Key GUWOSEVHCSNYFK-NSHDSACASA-N
Synonym inverted exclamation marka-cpsi 1306
PubChem CID 97992476
IUPAC Name 2-[(5S)-3-(2,4-difluorophenyl)-4,5-dihydro-1,2-oxazol-5-yl]-1-morpholin-4-ylethanone
SMILES C1COCCN1C(=O)CC2CC(=NO2)C3=C(C=C(C=C3)F)F

Specifications

Quantity 10 mg
Formula Weight 310.3
Percent Purity >98%
Chemical Name or Material (±)-CPSI 1306
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.