Learn More
Cytiva (Formerly Pall Lab) Pleated Capsule Filter with Versapor supported acrylic copolymer membrane, 0.2 m, sterile (1/pkg)
Supplier:  Cytiva 12117
Encompass
Designed both for prefiltration of particulate liquids (such as serum) prior to sterilizing filtration, and removal of debris and particles from liquids not requiring sterilization. Pleated Versapor supported acrylic copolymer membrane filter in a polycarbonate housing.-For prefiltration and removal of debris-Polycarbonate housing-Vented capsules This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3185