Learn More
Aviva Systems Biology Corp CPT1A Antibody - middle region (ARP44796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPT1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CPT1A. Alternate names: CPT1, CPT1-L, L-CPT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58894