Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CPT1A Antibody Blocking Peptide
SDP

Catalog No. NB153791PEP
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153791PEP 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153791PEP Supplier Novus Biologicals™ Supplier No. NB10053791PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from human CPT1A. Source: Synthetic Amino Acid Sequence: C-DPAQTVEQRLKLFK The CPT1A Blocking Peptide is derived from Synthetic. The CPT1A Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 1374
Protein Family CPT1A
Protein Subtype Blocking Peptide
Purification Method Immunogen affinity purified
Purity >50%, by HPLC
Common Name CPT1A Blocking Peptide
Concentration LYOPH
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt.
For Use With (Application) Antibody Competition
Gene Alias carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, Carnitine palmitoyltransferase 1A, carnitine palmitoyltransferase 1A (liver), carnitine palmitoyltransferase I, liver, CPT I, CPT1, CPT1-LEC 2.3.1.21, CPTI-L, EC 2.3.1, L-CPT1
Gene Symbol CPT1A
Label Type Unlabeled
Molecular Weight (g/mol) 88kDa
Product Type Peptide
Quantity 0.1 mg
Regulatory Status RUO
Research Category Neuroscience
Source Goat
Specific Reactivity Human
Immunogen C-DPAQTVEQRLKLFK
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.