Learn More
Cayman Chemical CPTH2 (hydrochloride), Purity: ≥95%, CAS: 2108899-91-6, 10 mg
SDP
Catalog No. 502954396
Encompass_Preferred
Specifically inhibits Gcn5-dependent acetylation of histone H3K14 at a concentration of 0.8 mM both in vitro and in vivo
Catalog No. 50-295-4396 Supplier Cayman Chemical Supplier No. 1208610 MG
Specifically inhibits Gcn5-dependent acetylation of histone H3K14 at a concentration of 0.8 mM both in vitro and in vivo