Learn More
Medchemexpress LLC CPTH6 hydrobromide 10mg | 2321332-57-2 | 386.74 g/mol | C15H17BrClN3S | 10 MG
SDP
Catalog No. 5000308276
Encompass_Preferred
CPTH6 hydrobromide is the hydrobromide salt of a thiazole-derived small molecule used to study histone acetylation, apoptosis, and autophagy in cancer cell models. It is supplied as a white to off-white solid with high reported purity and good solubility in polar solvents.
  • Inhibits histone acetyltransferase activity, reported for Gcn5 and pCAF.
  • Induces apoptotic and autophagic features in leukemia cell lines.
  • Supplied as a hydrobromide salt for improved handling and solubility.
  • High purity suitable for research applications (≥99.0%).
  • Soluble in DMSO (≈66.67 mg/mL) for in vitro assays.
  • Store sealed at -20°C; in solution, store at -80°C for long-term stability.
Catalog No. 50-003-08276 Supplier Medchemexpress LLC Supplier No. HY115406A10MG
CPTH6 hydrobromide is the hydrobromide salt of a thiazole-derived small molecule used to study histone acetylation, apoptosis, and autophagy in cancer cell models. It is supplied as a white to off-white solid with high reported purity and good solubility in polar solvents.
  • Inhibits histone acetyltransferase activity, reported for Gcn5 and pCAF.
  • Induces apoptotic and autophagic features in leukemia cell lines.
  • Supplied as a hydrobromide salt for improved handling and solubility.
  • High purity suitable for research applications (≥99.0%).
  • Soluble in DMSO (≈66.67 mg/mL) for in vitro assays.
  • Store sealed at -20°C; in solution, store at -80°C for long-term stability.