Learn More
Medchemexpress LLC CPTH6 hydrobromide 1mg | 2321332-57-2 | 1 MG
SDP
Catalog No. 5000228214
Encompass_Preferred
CPTH6 hydrobromide is a thiazole-derived small molecule that modulates histone acetyltransferase activity and induces apoptotic and autophagic features in leukemia cell lines. It is provided as the hydrobromide salt for use in biochemical and cell-based cancer research applications.
  • Thiazole-derived HAT inhibitor that reduces histone acetylation.
  • Promotes apoptotic program and increases autophagic features in leukemia cells.
  • Provided as a solid hydrobromide salt for improved handling and stability.
  • Suitable for biochemical assays and cell-based studies of acetylation-dependent processes.
  • Supplied in small research quantities for screening and mechanistic experiments.
Catalog No. 50-002-28214 Supplier Medchemexpress LLC Supplier No. HY115406A1MG
CPTH6 hydrobromide is a thiazole-derived small molecule that modulates histone acetyltransferase activity and induces apoptotic and autophagic features in leukemia cell lines. It is provided as the hydrobromide salt for use in biochemical and cell-based cancer research applications.
  • Thiazole-derived HAT inhibitor that reduces histone acetylation.
  • Promotes apoptotic program and increases autophagic features in leukemia cells.
  • Provided as a solid hydrobromide salt for improved handling and stability.
  • Suitable for biochemical assays and cell-based studies of acetylation-dependent processes.
  • Supplied in small research quantities for screening and mechanistic experiments.