Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CPTH6 hydrobromide 5mg | 2321332-57-2 | 5 MG
SDP

Catalog No. 5000308275
Encompass_Preferred

CPTH6 hydrobromide is the hydrobromide salt of CPTH6, a thiazole-derived inhibitor of histone acetyltransferases selective for Gcn5 and pCAF. It reduces acetylation of histone H3/H4 and α-tubulin, impairs autophagy, and induces G0/G1 arrest and apoptosis in tumor cell lines; it is used as a research reagent for epigenetics and cancer biology studies.

  • Research-grade compound for in vitro studies.
  • Purity ≥98%.
  • Molecular weight 386.7 g/mol.
  • Molecular formula C15H16ClN3S · HBr.
  • Soluble in DMSO, ethanol, and DMF.
  • Available in 5 mg vials.

Catalog No. 50-003-08275 Supplier Medchemexpress LLC Supplier No. HY115406A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CPTH6 hydrobromide is the hydrobromide salt of CPTH6, a thiazole-derived inhibitor of histone acetyltransferases selective for Gcn5 and pCAF. It reduces acetylation of histone H3/H4 and α-tubulin, impairs autophagy, and induces G0/G1 arrest and apoptosis in tumor cell lines; it is used as a research reagent for epigenetics and cancer biology studies.

  • Research-grade compound for in vitro studies.
  • Purity ≥98%.
  • Molecular weight 386.7 g/mol.
  • Molecular formula C15H16ClN3S · HBr.
  • Soluble in DMSO, ethanol, and DMF.
  • Available in 5 mg vials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.