Learn More
Cayman Chemical CPTH6hydrobromIde 10mg
SDP
Catalog No. 502984679
Encompass_Preferred
A thiazole derivative that inhibits the lysine acetyltransferase activity of Gcn5 and pCAF but not p300 or CBP; blocks the acetylation of H3/H4 histones and α-tubulin in several leukemia cell lines
Catalog No. 50-298-4679 Supplier Cayman Chemical Supplier No. 1982810 MG
A thiazole derivative that inhibits the lysine acetyltransferase activity of Gcn5 and pCAF but not p300 or CBP; blocks the acetylation of H3/H4 histones and α-tubulin in several leukemia cell lines