Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advanced ImmunoChemical, Inc. Recombinant Canine Parvovirus VP2 Protein, 1.0 mg, lyophilized, 584 amino acids, purity > 90%, expressed in insect cell line

Supplier:  Advanced ImmunoChemical, Inc. 8CPV21MG

Encompass_Preferred

This product is a recombinant canine parvovirus VP2 protein, essential for research and diagnostic applications related to CPV detection and antibody titre assessment. Expressed in an insect cell line, this protein is lyophilized for stability and ease of use, serving as a standard in immunoassays for CPV immunodetection and as an antigen for determining CPV antibody titres in canine blood. With a high purity level confirmed by SDS-PAGE and verified immunoreactivity with specific antibodies, it is reliable for various scientific applications.

  • Contains recombinant canine parvovirus VP2 capsid protein
  • 584 amino acid residues in length
  • Expressed in an insect cell line
  • Lyophilized presentation for stability
  • Purity verified to be greater than 90%
  • Applications include standardization in CPV immunoassays
  • Presented in a formulation compatible with various research needs
  • Storage recommended at -20°C

Catalog No. NC2078026


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.