Learn More
Adam Equipment CPWplus 75P - CPWplus Bench and Floor Scales - Capacity: 165lb / 75kg, Readability: 0.05lb / 0.02kg
Supplier:  Adam Equipment CPWPLUS 75P
Encompass_Preferred
The CPWplus Bench and Floor Scales from Adam Equipment offer a versatile, reliable solution for industries including food processing, shipping, materials testing, and veterinary use. Built with a grade 304 stainless steel pan and splashproof keypad, they withstand demanding environments.A bright backlit display with large digits ensures clear visibility, while the RS-232 interface enables easy data transfer to PCs and printers. The internal rechargeable battery and optional wall-mounted indicator provide flexibility for both portable and fixed applications. Multiple models—including compact, pillar-mounted, large platform, and mobile versions with wheels—meet a wide range of needs.Additional features such as color-coded keys, hold function, programmable backlight, auto power-off, digital filtering, and zero-tracking ensure efficient, accurate performance in any setting.
Catalog No. NC3964965