Learn More
MARSHALL SCIENTIFIC Branson CPX3800 Digital Ultrasonic Cleaner
SDP
Supplier:  MARSHALL SCIENTIFIC CPX952319R
Encompass_Preferred
Bransonic CPX ultrasonic baths feature digital controls with power tracking capabilities to adjust for light or heavy loads, high/low power control, degassing and sleep mode. The CPX baths offer the following features: Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance, even with multiple users or when bath conditions change. The CPX degassing period can be set up to 99 minutes. Extended degas time periods allow for “beyond cleaning” applications such as sample preparation, which includes degassing liquids, mixing and homogenization, dissolving solids, cell lysing, and dispersion of particles. When cleaning delicate parts, the CPX allows the user to lower the amplitude of the acoustic energy in the bath: 100% power to the tank for normal applications, 70% power for delicate applications. Manipulating the
Catalog No. NC2590758