Learn More
MARSHALL SCIENTIFIC Branson CPX5800H Digital Heated Ultrasonic Cleaner
SDP
Supplier:  MARSHALL SCIENTIFIC CPX952518R
Encompass_Preferred
Bransonic CPXH heated ultrasonic baths are the most technologically advanced ultrasonic baths available. With programmable features, digital operation with power tracking capabilities to adjust for light or heavy loads, high/low power control, and degassing, the CPXH is the most versatile system available. The CPXH degassing period can be set up to 99 minutes. Extended degas time periods allow for “beyond cleaning” applications such as sample preparation, which includes degassing liquids, mixing and homogenization, dissolving solids, cell lysing, and dispersion of particles. Constant activity/power tracking automatically maintains the same ultrasonic power by adjusting for changes in liquid level and temperature caused by a light or heavy load. This helps ensure more uniform cleaning and consistent performance, even with multiple users or when bath conditions change. Consult with your Branson representative about the Bransonic accessories and solution best suited for your application n
Catalog No. NC2590759