Learn More
Medchemexpress LLC Cq-lyso | 2089438-23-1 | 99.0% | 485.62 | C30H35N3O3 | 10 MG
SDP
Catalog No. 5000297854
Encompass_Preferred
CQ-Lyso is a lysosome-targeting chromenoquinoline fluorescent probe used to measure lysosomal pH in living cells with single-wavelength excitation. It enables visualization of dynamic pH changes in lysosomes and is supplied for research use only.
  • Lysosome-targeting chromenoquinoline fluorescent probe.
  • Measures lysosomal pH using single-wavelength excitation.
  • Enables visualization of dynamic pH changes in living cells.
  • High purity for reliable experimental results.
  • Soluble in DMSO and DMF at up to 100 mg/mL with warming and sonication.
  • Store solid at 4°C and protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research pack quantities (e.g., 10 mg).
Catalog No. 50-002-97854 Supplier Medchemexpress LLC Supplier No. HYD211910MG
Add to Cart
Add to Cart
CQ-Lyso is a lysosome-targeting chromenoquinoline fluorescent probe used to measure lysosomal pH in living cells with single-wavelength excitation. It enables visualization of dynamic pH changes in lysosomes and is supplied for research use only.
  • Lysosome-targeting chromenoquinoline fluorescent probe.
  • Measures lysosomal pH using single-wavelength excitation.
  • Enables visualization of dynamic pH changes in living cells.
  • High purity for reliable experimental results.
  • Soluble in DMSO and DMF at up to 100 mg/mL with warming and sonication.
  • Store solid at 4°C and protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research pack quantities (e.g., 10 mg).