Learn More
Adam Equipment CQT 2000 - Core Portable Compact Balances - Capacity: 2000g, Readability: 1g
Catalog No. NC3964743
Encompass_Preferred
Compact and durable, Core balances are the best choice for basic weighing and economy. The simple keypad with dual tare keys and a bright backlit display make Core balances easy to use. The tough durable ABS housing is designed to withstand field and laboratory environments, while being easy to clean and protected from accidental spills. With built-in ShockProtect overload protection Core balances can handle excessive overloads without a problem. Complete with a removable draft shield, AC adapter, and integral security slot, Core balances are ready to work right out of the box. Measures 6.7 inches wide x 9.6 deep x 3.1 tall.
Catalog No. NC3964743 Supplier Adam Equipment Supplier No. CQT 2000
Compact and durable, Core balances are the best choice for basic weighing and economy. The simple keypad with dual tare keys and a bright backlit display make Core balances easy to use. The tough durable ABS housing is designed to withstand field and laboratory environments, while being easy to clean and protected from accidental spills. With built-in ShockProtect overload protection Core balances can handle excessive overloads without a problem. Complete with a removable draft shield, AC adapter, and integral security slot, Core balances are ready to work right out of the box. Measures 6.7 inches wide x 9.6 deep x 3.1 tall.