Learn More
MSC Cestus Work Glove, 13g HPPE, Foam Nitrile, 1 ea.
Catalog No. 5000686682
Encompass
The Brutus LD is a cut resistant work glove designed for high-dexterity applications. Constructed with a 13 gauge HPPE knit, this form-fitting glove offers a reliable balance of protection and motor control. It features a foam nitrile coating for enhanced grip and is built with a reinforced saddle area between the thumb and index finger to improve durability in high-wear zones.
  • Features 13 gauge HPPE knit for cut resistance
  • Foam nitrile coating provides excellent grip
  • Reinforced saddle area with flat nitrile for added durability
  • Form-fitting design supports high dexterity and motor control
  • Compatible with touchscreen devices
Catalog No. 50-006-86682 Supplier MSC Supplier No. 12177069
Add to Cart
Add to Cart
The Brutus LD is a cut resistant work glove designed for high-dexterity applications. Constructed with a 13 gauge HPPE knit, this form-fitting glove offers a reliable balance of protection and motor control. It features a foam nitrile coating for enhanced grip and is built with a reinforced saddle area between the thumb and index finger to improve durability in high-wear zones.
  • Features 13 gauge HPPE knit for cut resistance
  • Foam nitrile coating provides excellent grip
  • Reinforced saddle area with flat nitrile for added durability
  • Form-fitting design supports high dexterity and motor control
  • Compatible with touchscreen devices