Learn More
Bevco Precision Manufacturing Co Dura Tall Height Heavy-Duty ISO 4 Cleanroom Black Polyurethane Chair; Non-Tilt; Polished Aluminum Base w/Adjustable Footirng
Catalog No. NC2023089 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
BEVCO's Dura heavy-duty ISO 4 Cleanroom polyurethane chairs are designed to meet the particulate goals of ISO 14644-1 and have been tested by an independent laboratory to certify ISO 4 compliance. The black polyurethane seat and back are impervious to water, oil, and most chemicals and are puncture resistant and easy to clean. This non-tilt model features manual back and seat height adjustments for increased comfort during extended sitting durations. The contoured back is fully adjustable encouraging proper spinal alignment. The large waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 20. 5" - 30. 5". This chair comes with a sturdy 27" diameter polished aluminum base with adjustable chrome footring. Model comes standard with mushroom glides. Optional casters can be ordered separately.
Catalog No. NC2023089 Supplier Bevco Precision Manufacturing Co Supplier No. 7550DC
BEVCO's Dura heavy-duty ISO 4 Cleanroom polyurethane chairs are designed to meet the particulate goals of ISO 14644-1 and have been tested by an independent laboratory to certify ISO 4 compliance. The black polyurethane seat and back are impervious to water, oil, and most chemicals and are puncture resistant and easy to clean. This non-tilt model features manual back and seat height adjustments for increased comfort during extended sitting durations. The contoured back is fully adjustable encouraging proper spinal alignment. The large waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 20. 5" - 30. 5". This chair comes with a sturdy 27" diameter polished aluminum base with adjustable chrome footring. Model comes standard with mushroom glides. Optional casters can be ordered separately.