Learn More
Uline Chemical Resistant Spray Bottle - 32 oz
Catalog No. NC2892839
Encompass
This item must be ordered in multiples of 3 per supplier requirements
Use with weed killers, pesticides and strippers. Super charged nozzle sprays three times more than standard nozzles. Graduated scale for easy mixing and diluting. Handy check-off grid to identify contents.
Catalog No. NC2892839 Supplier Uline Supplier No. S16187
This item must be ordered in multiples of 3 per supplier requirements
Use with weed killers, pesticides and strippers. Super charged nozzle sprays three times more than standard nozzles. Graduated scale for easy mixing and diluting. Handy check-off grid to identify contents.