Learn More
Julabo USA Inc 1m CR tubing (Chloroprene), 12mm (1/2") ID
SDP
Catalog No. 507534132
Encompass_Preferred
Bath Accessories, Julabo, CR tubing (chloroprene). Priced per 1m lengths. (-30 °C to +120 °C) 12 mm (1/2") inner diameter. Maximum pressure 1.2 bar / 17.4 PSI.
Catalog No. 50-753-4132 Supplier Julabo USA Inc Supplier No. 8930012
Bath Accessories, Julabo, CR tubing (chloroprene). Priced per 1m lengths. (-30 °C to +120 °C) 12 mm (1/2") inner diameter. Maximum pressure 1.2 bar / 17.4 PSI.