Learn More
Micronova™ CR100 PC™ Cleanroom Tape—Orange
SDP
Catalog No. 19035334G Shop All Micronova Manufacturing Products
Change view
Click to view available options
Width (English):
75 in.
Catalog No. Width (English)
19-035-334G 75 in.
1 options
Catalog No. 19-035-334G Supplier Micronova™ Supplier No. CR100PC34OR
Processed for applications in and around cleanrooms
  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag
Order Info
Available in red, yellow, white, black, brown, dark blue, dark green, light blue, light green, orange, purple, and transparent in 36 yd. long (33m) rolls. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here.
Specifications
Width (English) 75 in.