Learn More
RS AMERICAS INC CR2477N BATTERY COIN CELL
Catalog No. NC1067169
Encompass
Battery; Non-Rechargeable; Coin/Button; Lithium; 3VDC; 950mAh; SMT; Computer, 8.3g
Catalog No. NC1067169 Supplier RS AMERICAS INC Supplier No. 70157676
Battery; Non-Rechargeable; Coin/Button; Lithium; 3VDC; 950mAh; SMT; Computer, 8.3g