Learn More
Campbell Scientific CR6 Measurement and Control Datalogger
Catalog No. NC1790289
Encompass
The CR6 Measurement and Control Datalogger is a powerful core component for your data-acquisition system. The CR6 datalogger provides fast communication, low power requirements, built-in USB, compact size, and high analog input accuracy and resolution. It uses universal (U) terminals to allow a connection to virtually any sensor—analog, digital, or smart. This multipurpose data logger is also capable of doing static vibrating-wire measurements.
Catalog No. NC1790289 Supplier Campbell Scientific Supplier No. CR6NASTSWCC
The CR6 Measurement and Control Datalogger is a powerful core component for your data-acquisition system. The CR6 datalogger provides fast communication, low power requirements, built-in USB, compact size, and high analog input accuracy and resolution. It uses universal (U) terminals to allow a connection to virtually any sensor—analog, digital, or smart. This multipurpose data logger is also capable of doing static vibrating-wire measurements.