Learn More
TARGETMOL CHEMICALS INC CRA-026440 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581628
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CRA-026440 hydrochloride is a potent and broad-spectrum HDAC inhibitor. CRA-026440 hydrochloride exhibits Ki values against recombinant HDAC isoenzymes HDAC1 HDAC2 HDAC3 HDAC6 HDAC8 and HDAC10 are 4 nM 14 nM 11 nM 15 nM 7 nM and 20 nM respectively. CRA-026440 hydrochloride exhibits antitumor and antiangiogenic activities. purity: 99%
Catalog No. 50-258-1628 Supplier TARGETMOL CHEMICALS INC Supplier No. T10883L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CRA-026440 hydrochloride is a potent and broad-spectrum HDAC inhibitor. CRA-026440 hydrochloride exhibits Ki values against recombinant HDAC isoenzymes HDAC1 HDAC2 HDAC3 HDAC6 HDAC8 and HDAC10 are 4 nM 14 nM 11 nM 15 nM 7 nM and 20 nM respectively. CRA-026440 hydrochloride exhibits antitumor and antiangiogenic activities. purity: 99%