Learn More
Medchemexpress LLC CRA-026440 hydrochloride | 847459-98-7 | 98.1% | 456.92 | 1 ML
SDP
Catalog No. 5000402556
Encompass_Preferred
CRA-026440 hydrochloride is a potent, broad-spectrum Histone Deacetylase (HDAC) inhibitor. It exhibits antitumor and antiangiogenic activities. This compound also functions as a click chemistry reagent, containing an alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Potent, broad-spectrum HDAC inhibitor
  • Exhibits antitumor and antiangiogenic activities
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Shows antiproliferative effect on HUVEC endothelial cells
  • Inhibits tumor cell growth and induces apoptosis
  • Inhibits ex vivo angiogenesis
  • Reduces tumor growth in xenograft models
Catalog No. 50-004-02556 Supplier Medchemexpress LLC Supplier No. HY19754A10MM/1ML
CRA-026440 hydrochloride is a potent, broad-spectrum Histone Deacetylase (HDAC) inhibitor. It exhibits antitumor and antiangiogenic activities. This compound also functions as a click chemistry reagent, containing an alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Potent, broad-spectrum HDAC inhibitor
  • Exhibits antitumor and antiangiogenic activities
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Shows antiproliferative effect on HUVEC endothelial cells
  • Inhibits tumor cell growth and induces apoptosis
  • Inhibits ex vivo angiogenesis
  • Reduces tumor growth in xenograft models