Learn More
Medchemexpress LLC CRA-026440 hydrochloride | 847459-98-7 | 98.1% | 456.92 | 100 MG
SDP
Catalog No. 5000401070
Encompass_Preferred
CRA-026440 hydrochloride is a potent, broad-spectrum HDAC inhibitor. It shows antitumor and antiangiogenic activities. This compound is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, broad-spectrum HDAC inhibitor
  • Shows antitumor and antiangiogenic activities
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-004-01070 Supplier Medchemexpress LLC Supplier No. HY19754A100MG
CRA-026440 hydrochloride is a potent, broad-spectrum HDAC inhibitor. It shows antitumor and antiangiogenic activities. This compound is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, broad-spectrum HDAC inhibitor
  • Shows antitumor and antiangiogenic activities
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)