Learn More
Medchemexpress LLC CRA-2059 TFA | 99.9% | 864.83 | 1 ML
SDP
Catalog No. 5000385855
Encompass_Preferred
CRA-2059 is a highly specific and selective tryptase inhibitor, with a Ki of 620 pM for recombinant human tryptase-β (rHTβ). Tryptase is a trypsin-like serine protease found as a major protein component in human mast cell secretory granules. CRA-2059 has the potential for inflammatory bowel disease research.
  • Highly specific and selective tryptase inhibitor
  • Potential for inflammatory bowel disease research
  • Inhibits recombinant human tryptase-β (rHTβ) with a Ki of 620 pM
  • Targets tryptase, a trypsin-like serine protease
Catalog No. 50-003-85855 Supplier Medchemexpress LLC Supplier No. HY19303B10MM/1ML
CRA-2059 is a highly specific and selective tryptase inhibitor, with a Ki of 620 pM for recombinant human tryptase-β (rHTβ). Tryptase is a trypsin-like serine protease found as a major protein component in human mast cell secretory granules. CRA-2059 has the potential for inflammatory bowel disease research.
  • Highly specific and selective tryptase inhibitor
  • Potential for inflammatory bowel disease research
  • Inhibits recombinant human tryptase-β (rHTβ) with a Ki of 620 pM
  • Targets tryptase, a trypsin-like serine protease