Learn More
Aviva Systems Biology Corp CRABP2 Antibody - middle region (ARP41982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRABP2. It was validated on Western Blot. CRABP2,cellular retinoic acid binding protein 2,RBP6; CRABP-II; CRABP2,cellular retinoic acid-binding protein 2,cellular retinoic acid-binding protein 2,OTTHUMP00000038732,cellular retinoic acid-binding protein IIThe immunogen is a synthetic peptide directed towards the middle region of human CRABP2. Alternate names: CRABP-II, RBP6ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-970