Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CRABP2 protein, mouse (His) | >95.0% | 10 UG
SDP

Catalog No. 5000290139
Encompass_Preferred

Recombinant mouse CRABP2 (cellular retinoic acid-binding protein 2) is supplied as a His-tagged, partially truncated fragment (P2-E138) expressed in E. coli and provided lyophilized for biochemical and ligand-binding studies.

  • Expression system: E. coli.
  • Tag: N-terminal 6xHis.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Molecular weight: ~16 kDa.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Available sizes: 5 μg, 10 μg, 50 μg, 100 μg.
  • Storage: store at -20°C; aliquot and keep at -80°C for long term.
  • Reconstitution: recommended ≥100 μg/mL in ddH2O.

Catalog No. 50-002-90139 Supplier Medchemexpress LLC Supplier No. HYP7568910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse CRABP2 (cellular retinoic acid-binding protein 2) is supplied as a His-tagged, partially truncated fragment (P2-E138) expressed in E. coli and provided lyophilized for biochemical and ligand-binding studies.

  • Expression system: E. coli.
  • Tag: N-terminal 6xHis.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Molecular weight: ~16 kDa.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Available sizes: 5 μg, 10 μg, 50 μg, 100 μg.
  • Storage: store at -20°C; aliquot and keep at -80°C for long term.
  • Reconstitution: recommended ≥100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.