Learn More
Strem, An Ascensus Company CAS# 64536-78-3. 1g. (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I)hexafluorophosphate, 99% CRABTREE'S CATALYST. MFCD00075097
SDP
Catalog No. 502045397
Encompass
CAS# 64536-78-3. 1g. (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I)hexafluorophosphate, 99% CRABTREE'S CATALYST. MFCD00075097. Molecular Weight: 804.89. Molecular Formula: [(C6H11)3P][C8H12][C5H5N]Ir+PF6ae. Color/form: orange xtl. Strem# 77-9500. http://www.strem.com/catalog/v/77-9500/
Catalog No. 50-204-5397 Supplier Strem, An Ascensus Company Supplier No. 7795001G
CAS# 64536-78-3. 1g. (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I)hexafluorophosphate, 99% CRABTREE'S CATALYST. MFCD00075097. Molecular Weight: 804.89. Molecular Formula: [(C6H11)3P][C8H12][C5H5N]Ir+PF6ae. Color/form: orange xtl. Strem# 77-9500. http://www.strem.com/catalog/v/77-9500/