Learn More
Sino Biological CRACC / SLAM7 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11691MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CRACC / SLAM7 (rh CRACC / SLAM7; Catalog#11691-H08H; NP_067004.3; Met1-Met226). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8649