Learn More
Sino Biological CRACC / SLAM7 / CD319 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11691R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CRACC / SLAM7 / CD319 (rh CRACC / SLAM7 / CD319; Catalog#11691-H08H; NP_067004.3; Met1-Met226).
Catalog No. 50-224-7892