Learn More
Bellco Glass, Inc. Cradle Ring, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800453
Encompass_Preferred
Cradle Ring for Bottle Top Vacuum FiltrationThe Autofil hands-free filtration system works in conjunction with the Quick-Connect Pedestal or cradle ring. These two filtering options offer a permanent vacuum hose connection and minimize set-up time. The vacuum hose connects to the back of the pedestal or cradle ring and enables adjustable flow control using the on/off switch. This patented docking system is made from stainless steel.Allows for hands-free filtrationProvides on/off vacuum flow controlPermanent hose connection to minimize set-up timeWorks in conjunction with Autofil system in 15ml, 50ml, 250ml, 500ml, and 1000mlMade in the USASpecificationsHose Barb Dimensions: .38 inches, .45 inches and .5 inchesMaterial: SteelColor: Chrome
Catalog No. 50-255-0309